Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFA4L2
Entrez Gene ID
102179638
Gene Name
NDUFA4 mitochondrial complex associated like 2
Gene Aliases
-
Chromosome Location
Chr.5: 55533606 - 55535984 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048048.1 XP_017903537.1 3 - 4 324 73

View Full Product Details