Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IZUMO3
Entrez Gene ID
102171333
Gene Name
IZUMO family member 3
Gene Aliases
-
Chromosome Location
Chr.8: 19659798 - 19663008 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683598.3 XP_005683655.1 1 - 2 464 65
XM_005683599.2 XP_005683656.1 1 - 2 233 65

View Full Product Details