Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GZMK
Entrez Gene ID
106501752
Gene Name
granzyme K
Gene Aliases
-
Chromosome Location
Chr.20: 24125122 - 24135569 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972766.2 XP_013828220.1 4 - 5 695 63
GenBank mRNA JO608931.2 - 3 - 4 493 63

View Full Product Details