Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METRNL
Entrez Gene ID
102173882
Gene Name
meteorin like, glial cell differentiation regulator
Gene Aliases
-
Chromosome Location
Chr.19: 49192165 - 49207375 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065384.1 XP_017920873.1 3 - 4 840 77
GenBank mRNA KA313773.1 - 1 - 2 815 77

View Full Product Details