Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF11
Entrez Gene ID
102187777
Gene Name
Kruppel like factor 11
Gene Aliases
-
Chromosome Location
Chr.11: 87144508 - 87156011 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055668.1 XP_017911157.1 2 - 3 450 86
XM_018055669.1 XP_017911158.1 3 - 4 468 86
GenBank mRNA KA325641.1 - 2 - 3 343 86

View Full Product Details