Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PPP1R3F
Entrez Gene ID
102188204
Gene Name
protein phosphatase 1 regulatory subunit 3F
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017190157: 1999 - 19835 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700737.3 XP_005700794.3 1 - 2 4068 87
XM_013976470.2 XP_013831924.2 1 - 2 4068 87
GenBank mRNA JO639149.2 - 3 - 4 1893 87

View Full Product Details