The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018056524.1 | XP_017912013.1 | 5 - 6 | 1043 | 89 |
XM_018056525.1 | XP_017912014.1 | 5 - 6 | 1063 | 89 | |
XM_018056526.1 | XP_017912015.1 | 5 - 6 | 1175 | 89 |