Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM71E1
Entrez Gene ID
102189894
Gene Name
family with sequence similarity 71 member E1
Gene Aliases
-
Chromosome Location
Chr.18: 57622155 - 57628104 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692863.3 XP_005692920.3 3 - 4 496 66
XM_018062886.1 XP_017918375.1 3 - 4 532 66
GenBank mRNA KA333992.1 - 1 - 2 80 66

View Full Product Details