Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK2
Entrez Gene ID
100860751
Gene Name
cyclin dependent kinase 2
Gene Aliases
-
Chromosome Location
Chr.5: 56713350 - 56718919 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285608.1 NP_001272537.1 2 - 3 366 103
XM_013963900.1 XP_013819354.1 2 - 3 431 103
GenBank mRNA EF035041.1 - 2 - 3 366 103

View Full Product Details