Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP3
Entrez Gene ID
100860846
Gene Name
fatty acid binding protein 3
Gene Aliases
H-FABP
Chromosome Location
Chr.2: 13664759 - 13673331 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285701.1 NP_001272630.1 2 - 3 245 58
XM_013971605.2 XP_013827059.1 3 - 4 438 58
GenBank mRNA AY466498.1 - 2 - 3 248 58
FJ844408.1 - 2 - 3 247 58
HQ404383.1 - 2 - 3 244 58
HQ738535.1 - 2 - 3 244 58
HQ895788.1 - 2 - 3 244 58
JQ031287.1 - 2 - 3 246 58

View Full Product Details