Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OGN
Entrez Gene ID
100860756
Gene Name
osteoglycin
Gene Aliases
-
Chromosome Location
Chr.8: 83789288 - 83805336 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285613.1 NP_001272542.1 5 - 6 723 102
XM_013966068.2 XP_013821522.2 6 - 7 1289 102
XM_018051762.1 XP_017907251.1 6 - 7 997 102
GenBank mRNA EF151287.1 - 5 - 6 723 102

View Full Product Details