Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLIRP
Entrez Gene ID
102173011
Gene Name
SRA stem-loop interacting RNA binding protein
Gene Aliases
-
Chromosome Location
Chr.10: 14239464 - 14245317 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686168.3 XP_005686225.2 3 - 4 299 98
GenBank mRNA JR119187.1 - - 2337 98

View Full Product Details