Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC10A5
Entrez Gene ID
102175597
Gene Name
solute carrier family 10 member 5
Gene Aliases
-
Chromosome Location
Chr.14: 1327220 - 1334971 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689278.3 XP_005689335.1 1 - 1 2101 97

View Full Product Details