Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CX3CL1
Entrez Gene ID
102189703
Gene Name
C-X3-C motif chemokine ligand 1
Gene Aliases
-
Chromosome Location
Chr.18: 26740198 - 26752245 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063153.1 XP_017918642.1 1 - 2 212 81

View Full Product Details