Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNIP1
Entrez Gene ID
102175084
Gene Name
Smad nuclear interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 12453765 - 12469773 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678684.3 XP_005678741.1 3 - 4 995 64
GenBank mRNA JO342636.2 - 1 - 2 474 64
KA318601.1 - 1 - 2 678 64

View Full Product Details