Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC8
Entrez Gene ID
102173225
Gene Name
fibronectin type III domain containing 8
Gene Aliases
-
Chromosome Location
Chr.19: 14603775 - 14615608 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693207.2 XP_005693264.1 2 - 3 640 94

View Full Product Details