Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2D4
Entrez Gene ID
102168669
Gene Name
ubiquitin conjugating enzyme E2 D4 (putative)
Gene Aliases
-
Chromosome Location
Chr.22: 246931 - 272838 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695485.3 XP_005695542.1 2 - 3 201 109
XM_005695486.3 XP_005695543.1 2 - 3 200 109
XM_018038281.1 XP_017893770.1 2 - 3 201 109
XM_018038282.1 XP_017893771.1 2 - 3 201 109
XM_018038283.1 XP_017893772.1 2 - 3 201 109
GenBank mRNA JO605864.2 - 7 - 8 615 109

View Full Product Details