Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A18
Entrez Gene ID
102175556
Gene Name
membrane spanning 4-domains A18
Gene Aliases
-
Chromosome Location
Chr.29: 36334769 - 36347897 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042811.1 XP_017898300.1 5 - 6 852 67

View Full Product Details