Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCL2
Entrez Gene ID
102181108
Gene Name
phosducin like 2
Gene Aliases
-
Chromosome Location
Chr.6: 71556442 - 71578984 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964523.1 XP_013819977.1 4 - 5 560 104

View Full Product Details