Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIPIN
Entrez Gene ID
102189311
Gene Name
TIMELESS interacting protein
Gene Aliases
-
Chromosome Location
Chr.10: 88019120 - 88035931 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053621.1 XP_017909110.1 4 - 5 396 90
XM_018053622.1 XP_017909111.1 5 - 6 406 90
XM_018053623.1 XP_017909112.1 4 - 5 473 90
XM_018053624.1 XP_017909113.1 3 - 4 246 90
GenBank mRNA JO339935.2 - 2 - 3 169 90
KA333207.1 - 4 - 5 377 90

View Full Product Details