Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT6
Entrez Gene ID
102190837
Gene Name
synaptotagmin 6
Gene Aliases
-
Chromosome Location
Chr.3: 91304620 - 91371432 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045940.1 XP_017901429.1 4 - 5 1615 64
XM_018045941.1 XP_017901430.1 4 - 5 1615 64

View Full Product Details