Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC10
Entrez Gene ID
102170117
Gene Name
leucine rich repeat containing 10
Gene Aliases
-
Chromosome Location
Chr.5: 43346608 - 43347665 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680186.3 XP_005680243.2 1 - 1 795 97
GenBank mRNA JO381085.2 - 1 - 1 434 97

View Full Product Details