Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VXN
Entrez Gene ID
102168364
Gene Name
vexin
Gene Aliases
C14H8orf46
Chromosome Location
Chr.14: 51341151 - 51367395 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058448.1 XP_017913937.1 1 - 2 489 103
GenBank mRNA JO636843.2 - 1 - 2 304 103

View Full Product Details