Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G2C
Entrez Gene ID
102168220
Gene Name
phospholipase A2 group IIC
Gene Aliases
-
Chromosome Location
Chr.2: 3481211 - 3507659 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676905.3 XP_005676962.2 1 - 2 389 90
GenBank mRNA JO434016.2 - 1 - 2 248 90

View Full Product Details