Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS35
Entrez Gene ID
102172748
Gene Name
VPS35 retromer complex component
Gene Aliases
-
Chromosome Location
Chr.18: 22271151 - 22300389 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691946.3 XP_005692003.1 9 - 10 1077 118
GenBank mRNA JO426407.1 - 8 - 9 2098 118

View Full Product Details