Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZAR1
Entrez Gene ID
102178614
Gene Name
zygote arrest 1
Gene Aliases
-
Chromosome Location
Chr.6: 67713658 - 67730970 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964626.2 XP_013820080.2 2 - 3 947 84

View Full Product Details