Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIPI
Entrez Gene ID
102183181
Gene Name
lipase I
Gene Aliases
-
Chromosome Location
Chr.1: 21585460 - 21645951 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049348.1 XP_017904837.1 3 - 4 695 90

View Full Product Details