Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNPLA1
Entrez Gene ID
102189446
Gene Name
patatin like phospholipase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.23: 38485370 - 38538191 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973931.2 XP_013829385.1 6 - 7 1865 71
XM_018039266.1 XP_017894755.1 6 - 7 1862 71
XM_018039267.1 XP_017894756.1 6 - 7 1866 71
GenBank mRNA KA327333.1 - 6 - 7 1903 71

View Full Product Details