Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYPC
Entrez Gene ID
102187901
Gene Name
glycophorin C (Gerbich blood group)
Gene Aliases
-
Chromosome Location
Chr.2: 57063316 - 57110274 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676274.3 XP_005676331.1 2 - 3 466 60

View Full Product Details