Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM46
Entrez Gene ID
102183088
Gene Name
RNA binding motif protein 46
Gene Aliases
-
Chromosome Location
Chr.17: 68298889 - 68341986 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061628.1 XP_017917117.1 1 - 2 154 69

View Full Product Details