Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOK2
Entrez Gene ID
102168348
Gene Name
docking protein 2
Gene Aliases
-
Chromosome Location
Chr.8: 68896446 - 68901071 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683968.3 XP_005684025.2 4 - 5 718 79
GenBank mRNA JO595694.2 - 4 - 5 759 79
JO595696.2 - 3 - 4 1743 79

View Full Product Details