Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHBDD2
Entrez Gene ID
102173907
Gene Name
rhomboid domain containing 2
Gene Aliases
-
Chromosome Location
Chr.25: 33908098 - 33916220 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040863.1 XP_017896352.1 4 - 5 876 81

View Full Product Details