The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018042501.1 | XP_017897990.1 | 2 - 3 | 377 | 110 |
XM_018042502.1 | XP_017897991.1 | 3 - 4 | 426 | 110 | |
GenBank mRNA | KA315623.1 | - | 2 - 3 | 410 | 110 |