Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YTHDF2
Entrez Gene ID
102177573
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Gene Aliases
-
Chromosome Location
Chr.2: 11217601 - 11248111 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057048.1 XP_017912537.1 4 - 5 2227 70
GenBank mRNA JT672185.1 - 1 - 2 175 70

View Full Product Details