Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CARNS1
Entrez Gene ID
102182385
Gene Name
carnosine synthase 1
Gene Aliases
-
Chromosome Location
Chr.29: 42768728 - 42777624 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042905.1 XP_017898394.1 4 - 5 466 102
XM_018042906.1 XP_017898395.1 3 - 4 192 102
GenBank mRNA JO636404.2 - 6 - 7 2796 102

View Full Product Details