Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
FRMD1
Entrez Gene ID
106502497
Gene Name
FERM domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189858: 7106 - 18923 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044881.1 XP_017900370.1 3 - 4 520 73
XM_018044882.1 XP_017900371.1 2 - 3 654 73
XM_018044883.1 XP_017900372.1 3 - 4 520 73
XM_018044884.1 XP_017900373.1 3 - 4 520 73
XM_018044885.1 XP_017900374.1 3 - 4 520 73
XM_018044886.1 XP_017900375.1 3 - 4 520 73
GenBank mRNA JO428985.2 - 2 - 3 126 73

View Full Product Details