Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEPE
Entrez Gene ID
102175196
Gene Name
matrix extracellular phosphoglycoprotein
Gene Aliases
-
Chromosome Location
Chr.6: 37364321 - 37378676 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681451.3 XP_005681508.2 4 - 5 226 106

View Full Product Details