Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA9
Entrez Gene ID
102182305
Gene Name
spermatogenesis associated 9
Gene Aliases
-
Chromosome Location
Chr.7: 15554155 - 15584326 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050025.1 XP_017905514.1 4 - 5 577 81

View Full Product Details