Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL37
Entrez Gene ID
102179292
Gene Name
interleukin 37
Gene Aliases
-
Chromosome Location
Chr.11: 46246080 - 46257477 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KA329633.1 - 4 - 5 852 79

View Full Product Details