Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK11
Entrez Gene ID
102179409
Gene Name
kallikrein related peptidase 11
Gene Aliases
-
Chromosome Location
Chr.18: 58093459 - 58098320 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062901.1 XP_017918390.1 6 - 7 905 72

View Full Product Details