Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRM2
Entrez Gene ID
102170576
Gene Name
mitochondrial rRNA methyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.25: 41814358 - 41823171 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040286.1 XP_017895775.1 2 - 3 363 62

View Full Product Details