Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD9
Entrez Gene ID
102191010
Gene Name
PDZ domain containing 9
Gene Aliases
-
Chromosome Location
Chr.25: 19625147 - 19644111 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040736.1 XP_017896225.1 4 - 5 894 102

View Full Product Details