Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PTMS
Entrez Gene ID
106502142
Gene Name
parathymosin
Gene Aliases
-
Chromosome Location
Chr.5: 102597980 - 102602829 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048558.1 XP_017904047.1 4 - 5 689 73
GenBank mRNA JO338849.2 - 4 - 5 288 73

View Full Product Details