Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNE4
Entrez Gene ID
102185961
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 4
Gene Aliases
-
Chromosome Location
Chr.2: 24588982 - 24592279 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676611.3 XP_005676668.1 1 - 2 198 81

View Full Product Details