Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFR1
Entrez Gene ID
102188281
Gene Name
SWI5 dependent homologous recombination repair protein 1
Gene Aliases
-
Chromosome Location
Chr.26: 27066272 - 27070225 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698434.3 XP_005698491.1 3 - 4 640 86
XM_005698436.3 XP_005698493.1 3 - 4 642 86
XM_018041679.1 XP_017897168.1 3 - 4 620 86

View Full Product Details