Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU4F3
Entrez Gene ID
102169585
Gene Name
POU class 4 homeobox 3
Gene Aliases
-
Chromosome Location
Chr.7: 52932347 - 52933837 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051723.1 XP_017907212.1 1 - 2 202 69

View Full Product Details