Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EOMES
Entrez Gene ID
102173338
Gene Name
eomesodermin
Gene Aliases
-
Chromosome Location
Chr.22: 2005808 - 2012330 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695502.3 XP_005695559.2 5 - 6 1787 72

View Full Product Details