Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC108636028
Entrez Gene ID
108636028
Gene Name
chymotrypsinogen A-like
Gene Aliases
-
Chromosome Location
Chr.5: 78736094 - 78754435 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048364.1 XP_017903853.1 1 - 2 264 108

View Full Product Details