Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZRSR2
Entrez Gene ID
102178809
Gene Name
zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 54473970 - 54497021 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005701066.3 XP_005701123.3 2 - 3 154 59

View Full Product Details