Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZRSR2
- Entrez Gene ID
-
102178809
- Gene Name
- zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X|NW_017189516: 54473970 - 54497021 on Build ARS1
- Gene Symbol
- LOC108634621
- Entrez Gene ID
-
108634621
- Gene Name
- U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Un|NW_017189685: 333142 - 374975 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO371925.2 |
- |
2 - 3 |
156 |
67 |
View Full Product Details